Invitrogen
Manufacturer Code:PA525583
Catalog # PIPA525583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 397-425 amino acids from the central region of human MCCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-methylcrotonyl-CoA carboxylase 1; 3-methylcrotonyl-CoA carboxylase biotin-containing subunit; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha; MCC-B MCCA 3-methylcrotonyl-CoA carboxylase 1 3-methylcrotonyl-CoA carboxylase biotin-containing subunit 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha MCCase subunit alpha methylcrotonoyl-CoA carboxylase subunit alpha mitochondrial methylcrotonoyl-Coenzy; MCCase subunit alpha; methylcrotonoyl-CoA carboxylase 1 (alpha); methylcrotonoyl-CoA carboxylase alpha; Methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial; methylcrotonoyl-Coenzyme A carboxylase 1 (alpha)
Gene Aliases: 1810045E08Rik; 2310058B18Rik; MCC-B; MCCA; MCCalpha; MCCC1; R75106
UniProt ID: (Human) Q96RQ3, (Mouse) Q99MR8
Entrez Gene ID: (Human) 56922, (Mouse) 72039
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.