Novus Biologicals
Manufacturer Code:NBP287786
Catalog # NB035408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human MCCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-methylcrotonyl-CoA carboxylase 1; 3-methylcrotonyl-CoA carboxylase 1 DKFZp686B20267 EC 6.4.1 EC 6.4.1.4 MCCAFLJ25545 MCC-B methylcrotonoyl-CoA carboxylase 1 (alpha) methylcrotonoyl-Coenzyme A carboxylase 1 (alpha) mitochondrial; 3-methylcrotonyl-CoA carboxylase biotin-containing subunit; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha; MCCase subunit alpha; methylcrotonoyl-CoA carboxylase 1 (alpha); methylcrotonoyl-CoA carboxylase alpha; Methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial; methylcrotonoyl-Coenzyme A carboxylase 1 (alpha)
Gene Aliases: MCC-B; MCCA; MCCC1
UniProt ID: (Human) Q96RQ3
Entrez Gene ID: (Human) 56922
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.