Proteintech
Manufacturer Code:148611AP150UL
Catalog # 50-172-9784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MCCC1 Fusion Protein Ag6663 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-methylcrotonyl-CoA carboxylase 1; 3-methylcrotonyl-CoA carboxylase 1 {ECO:0000250|UniProtKB:Q96RQ3}; 3-methylcrotonyl-CoA carboxylase biotin-containing subunit; 3-methylcrotonyl-CoA carboxylase biotin-containing subunit {ECO:0000250|UniProtKB:Q96RQ3}; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha {ECO:0000250|UniProtKB:Q96RQ3}; MCC B MCCA MCCase subunit alpha MCCC1; MCCase subunit alpha; MCCase subunit alpha {ECO:0000250|UniProtKB:Q96RQ3}; mccc1 {ECO:0000312|EMBL:AAH88473.1, ECO:0000312|RGD:1310615}; methylcrotonoyl-CoA carboxylase 1 (alpha); methylcrotonoyl-CoA carboxylase alpha; Methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial; methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial {ECO:0000250|UniProtKB:Q96RQ3}; methylcrotonoyl-Coenzyme A carboxylase 1 (alpha)
Gene Aliases: 1810045E08Rik; 2310058B18Rik; MCC-B; MCCA; MCCalpha; MCCC1; R75106
UniProt ID: (Human) Q96RQ3, (Rat) Q5I0C3, (Mouse) Q99MR8
Entrez Gene ID: (Human) 56922, (Rat) 294972, (Mouse) 72039
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.