Southern Biotechnology
Manufacturer Code:838509
Catalog # OB838509
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Chicken |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | C264 |
Immunogen | Mixture of E15 and E16 thymocytes from H.B15 H.B19 and H.B21 chicken embryos |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C Protect from light Do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.