Invitrogen
Manufacturer Code:PA577489
Catalog # PIPA577489
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RYNSVRQRTSMKG corresponding to amino acid residues 224-236 of mouse Melanocortin Receptor 5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MC5-R; MC5R; Melanocortin receptor 5
Gene Aliases: Mc5r
UniProt ID: (Mouse) P41149, (Rat) P35345
Entrez Gene ID: (Mouse) 17203, (Rat) 25726
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.