Invitrogen
Manufacturer Code:PA575353
Catalog # PIPA575353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids C-terminus of Human MC5R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MC-2; MC5-R; Melanocortin receptor 5; Melanocortin receptor 5 MC5-R MC-2 MC5R; melanocortin-5 receptor
Gene Aliases: MC2; MC5R
UniProt ID: (Human) P33032, (Mouse) P41149, (Rat) P35345
Entrez Gene ID: (Human) 4161, (Mouse) 17203, (Rat) 25726
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.