Invitrogen
Manufacturer Code:PA533929
Catalog # PIPA533929
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from 1st extracellular domain of human MC5 Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MC-2; MC5-R; MC5R HMC5R MC2 MC5-R Melanocortin 5 receptor MC5 MC5 receptor MC-2 Melanocortin receptor 5 Melanocortin-5 receptor; Melanocortin receptor 5; melanocortin-5 receptor
Gene Aliases: MC2; MC5R
UniProt ID: (Human) P33032
Entrez Gene ID: (Human) 4161
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.