Invitrogen
Manufacturer Code:PA572246
Catalog # PIPA572246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 25-57 amino acids from the human region of DANRE mc4r |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MC4-R; MC4R Melanocortin receptor 4 MC4-R MC4 MC4 receptor Melanocortin 4 receptor; Melanocortin receptor 4
Gene Aliases: MC4-R; mc4r; si:dkey-151m6.62; zMC4
UniProt ID: (Zebrafish) B0V1P1
Entrez Gene ID: (Zebrafish) 286833
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.