Invitrogen
Manufacturer Code:PA577488
Catalog # PIPA577488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)NSDSLTLEDQFIQHMD corresponding to amino acid residues 102-117 of rat MC3R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MC3-R; MC3R; Melanocortin receptor 3; obesity quantitative trait locus
Gene Aliases: BMIQ9; MC3; MC3-R; MC3R; OB20; OQTL
UniProt ID: (Human) P41968, (Mouse) P33033, (Rat) P32244
Entrez Gene ID: (Human) 4159, (Mouse) 17201, (Rat) 29310
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.