Invitrogen
Manufacturer Code:PA595419
Catalog # PIPA595419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human MC2 receptor different from the related mouse sequence by four amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACTH receptor; Adrenocorticotropic hormone receptor; Adrenocorticotropic hormone receptor ACTH receptor ACTH-R Adrenocorticotropin receptor Melanocortin receptor 2 MC2-R MC2R ACTHR; Adrenocorticotropin receptor; corticotropin receptor; MC2 receptor; MC2-R; melanocortin 2 receptor (adrenocorticotropic hormone); Melanocortin receptor 2
Gene Aliases: ACTH-R; ACTHR; MC2-R; MC2R
UniProt ID: (Human) Q01718, (Mouse) Q64326
Entrez Gene ID: (Human) 4158, (Mouse) 17200
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.