Invitrogen
Manufacturer Code:PA518867
Catalog # PIPA518867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (ENINNTARNNSDC) corresponding to the N-terminus amino acids of MC2R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACTH receptor; Adrenocorticotropic hormone receptor; Adrenocorticotropin receptor; corticotropin receptor; MC2 receptor; MC2-R; MC2R ACTHR melanocortin 2 receptor (adrenocorticotropic hormone) MGC125798 ACTH receptor MC2 receptor adrenocorticotropic hormone receptor corticotropin receptor melanocortin 2 receptor; melanocortin 2 receptor (adrenocorticotropic hormone); Melanocortin receptor 2
Gene Aliases: ACTHR; MC2R
UniProt ID: (Human) Q01718
Entrez Gene ID: (Human) 4158
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.