Invitrogen
Manufacturer Code:MA537591
Catalog # PIMA537591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 578CT6.2.3 |
Immunogen | KLH conjugated synthetic peptide between 205-232 amino acids from the Central region of human MC1R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha melanocyte stimulating hormone receptor; alpha melanocyte stimulating hormone receptor CMM5 e extension recessive yellow MC1 receptor Mc1r MC1-R melanocortin 1 receptor melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor) melanocortin receptor 1 melanocortin-1 receptor melanocyte-stimulating hormone receptor melanotropin receptor MSHR MSH-R Mshra SHEP2 Tob tobacco darkening; MC1-R; melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor); Melanocortin receptor 1; Melanocyte-stimulating hormone receptor; melanotropin receptor; MSH-R
Gene Aliases: CMM5; MC1R; MSH-R; MSHR; SHEP2
UniProt ID: (Human) Q01726
Entrez Gene ID: (Human) 4157
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.