Invitrogen
Manufacturer Code:PA595418
Catalog # PIPA595418
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human MC1 Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha melanocyte stimulating hormone receptor; extension recessive yellow; MC1-R; melanocortin 1 receptor; melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor); Melanocortin receptor 1; melanocortin-1 receptor; Melanocyte-stimulating hormone receptor; Melanocyte-stimulating hormone receptor MSH-R Melanocortin receptor 1 MC1-R MC1R MSHR; melanotropin receptor; MSH-R; tobacco darkening
Gene Aliases: CMM5; e; MC1R; MSH-R; MSHR; Mshra; SHEP2; Tob
UniProt ID: (Human) Q01726, (Mouse) Q01727
Entrez Gene ID: (Human) 4157, (Rat) 102552838, (Mouse) 17199
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.