Abnova
Manufacturer Code:MAB0001
Catalog # 89-107-813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AF-20 |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T696 of human MBS/MYPT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic phosphopeptide of MBS/MYPT1.
For Research Use Only
Protein Aliases: MBS; MGC133042; myosin binding subunit; Myosin phosphatase target subunit 1; myosin phosphatase, target subunit 1; Myosin phosphatase-targeting subunit 1; MYPT1; Protein phosphatase 1 regulatory subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory subunit 12A; Protein phosphatase myosin-binding subunit
Gene Aliases: M130; MBS; MYPT1; PPP1R12A
UniProt ID: (Human) O14974
Entrez Gene ID: (Human) 4659
Molecular Function: enzyme modulator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.