Novus Biologicals
Manufacturer Code:NBP1052030.025ML
Catalog # NB018356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7G7 |
Immunogen | Purified myelin basic protein isolated from bovine brain epitope is in peptide TPPPSQGKG amino acids 125-133 of the human 21.5kDa sequence. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MBP; MGC99675 Myelin A1 protein myelin basic protein Myelin membrane encephalitogenic protein; Myelin A1 protein; Myelin basic protein; Myelin membrane encephalitogenic protein
Gene Aliases: MBP
UniProt ID: (Human) P02686
Entrez Gene ID: (Human) 4155
Molecular Function: myelin protein structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.