Invitrogen
Manufacturer Code:PA578397
Catalog # PIPA578397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse Myelin Basic Protein. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C76307 Hmbpr Mbp Myelin Basic Protein Myelin Basic Protein R75289 golli mbp mld shi; Golli-Mbp; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; MBP; MBP S; Myelin A1 protein; Myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; shiverer
Gene Aliases: C76307; golli-mbp; Hmbpr; Mbp; Mbps; mld; R75289; Shi
UniProt ID: (Rat) P02688, (Mouse) P04370
Entrez Gene ID: (Rat) 24547, (Mouse) 17196
Molecular Function:
myelin protein
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.