Invitrogen
Manufacturer Code:PA543430
Catalog # PIPA543430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human LENG4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerophosphatidylinositol O-acyltransferase; Bladder and breast carcinoma-overexpressed gene 1 protein; h-mboa-7; leukocyte receptor cluster (LRC) member 4; Leukocyte receptor cluster member 4; LPIAT; LPIAT1; LPLAT 7; lyso-PI acyltransferase; Lysophosphatidylinositol acyltransferase; Lysophosphatidylinositol acyltransferase 1; Lysophospholipid acyltransferase 7; m-mboa-7; malignant cell expression-enhanced gene/tumor progression-enhanced; Membrane-bound O-acyltransferase domain-containing protein 7; O-acyltransferase domain-containing protein 7
Gene Aliases: 5730589L02Rik; BB1; hMBOA-7; LENG4; LPIAT; Lpiat1; LRC4; mBB1; MBOA7; MBOAT7; OACT7; RGD1306945
UniProt ID: (Bovine) Q0VCY6, (Human) Q96N66, (Mouse) Q8CHK3
Entrez Gene ID: (Bovine) 504236, (Horse) 100052743, (Dog) 100683790, (Human) 79143, (Rat) 308309, (Guinea pig) 100735624, (Mouse) 77582
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.