Abnova
Manufacturer Code:H00079143K
Catalog # 89-204-261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MBOAT7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MBOAT7 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 1-acylglycerophosphatidylinositol O-acyltransferase; BB1 LENG4 LPIAT hMBOA-7; Bladder and breast carcinoma-overexpressed gene 1 protein; h-mboa-7; leukocyte receptor cluster (LRC) member 4; Leukocyte receptor cluster member 4; LPIAT; LPLAT 7; lyso-PI acyltransferase; Lysophosphatidylinositol acyltransferase; Lysophospholipid acyltransferase 7; malignant cell expression-enhanced gene/tumor progression-enhanced; Membrane-bound O-acyltransferase domain-containing protein 7; O-acyltransferase domain-containing protein 7
Gene Aliases: BB1; hMBOA-7; LENG4; LPIAT; LRC4; MBOA7; MBOAT7; OACT7
UniProt ID: (Human) Q96N66
Entrez Gene ID: (Human) 79143
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.