Invitrogen
Manufacturer Code:PA532916
Catalog # PIPA532916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 14 amino acid peptide from internal region of human MBOAT4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ghrelin O-acyltransferase; GOAT OACT4 O-acyltransferase (membrane bound) domain containing 4 O-acyltransferase domain-containing protein 4 ghrelin O-acyltransferase membrane-bound O-acyltransferase domain-containing protein 4; Membrane-bound O-acyltransferase domain-containing protein 4; O-acyltransferase (membrane bound) domain containing 4; O-acyltransferase domain-containing protein 4
Gene Aliases: FKSG89; GOAT; MBOAT4; OACT4
UniProt ID: (Human) Q96T53
Entrez Gene ID: (Human) 619373
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.