Invitrogen
Manufacturer Code:PA553238
Catalog # PIPA553238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MBOAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerophosphate O-acyltransferase; 1-acylglycerophosphate O-acyltransferase MBOAT2; 1-acylglycerophosphocholine O-acyltransferase MBOAT2; 1-acylglycerophosphoethanolamine MBOAT2 O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase; LPAAT; LPCAT; LPEAT; LPLAT 2; lyso-PA acyltransferase; Lyso-PC acyltransferase 4; lyso-PE acyltransferase; Lysophosphatidic acid acyltransferase; Lysophosphatidylcholine acyltransferase; Lysophosphatidylcholine acyltransferase 4; Lysophosphatidylethanolamine acyltransferase; Lysophospholipid acyltransferase 2; Lysophospholipid acyltransferase 2 LPLAT 2 231- 1-acylglycerophosphate O-acyltransferase 1-acylglycerophosphoethanolamine O-acyltransferase 231n7 Lysophosphatidic acid acyltransferase LPAAT Lyso-PA acyltransferase Lysophosphatidylethanolamine acyltransferase LPEAT Lyso-PE acyltransferase Membrane-bound O-acyltransferase domain-containing protein 2 O-acyltransferase domain-containing protein 2 MBOAT2 OACT2; Membrane-bound O-acyltransferase domain-containing protein 2; O-acyltransferase (membrane bound) domain containing 2; O-acyltransferase domain-containing protein 2
Gene Aliases: LPAAT; LPCAT4; LPEAT; LPLAT 2; MBOAT2; OACT2
UniProt ID: (Human) Q6ZWT7
Entrez Gene ID: (Human) 129642
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.