Invitrogen
Manufacturer Code:PA543193
Catalog # PIPA543193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MBOAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase MBOAT1; 1-acylglycerophosphoserine O-acyltransferase; 1-acylglycerophosphoserine O-acyltransferase MBOAT1; LPE acyltransferase 1; LPEAT; LPLAT 1; LPSAT; lyso-PE acyltransferase; lyso-PS acyltransferase; lysophosphatidylethanolamine acyltransferase; Lysophosphatidylethanolamine acyltransferase 1; Lysophosphatidylserine acyltransferase; Lysophospholipid acyltransferase 1; membrane bound O-acyl transferase family, member 1; Membrane bound O-acyltransferase domain containing 1 MGC44669 OACT1 dJ434O11.1 LPEAT1; Membrane-bound O-acyltransferase domain-containing protein 1; O-acyltransferase (membrane bound) domain containing 1; O-acyltransferase domain-containing protein 1
Gene Aliases: 9130215M02Rik; BC023845; dJ434O11.1; LPEAT1; LPLAT; LPLAT 1; LPSAT; MBOAT1; Moact1; OACT1; RGD1565521; RGD1565561
UniProt ID: (Human) Q6ZNC8, (Mouse) Q8BH98
Entrez Gene ID: (Pig) 100152017, (Rabbit) 100353609, (Horse) 100066632, (Human) 154141, (Rat) 498741, (Mouse) 218121
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.