Invitrogen
Manufacturer Code:PA579640
Catalog # PIPA579640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat MBL2 recombit protein (Position: E19-D244). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2 (protein C); mannose binding lectin, liver (C); Mannose-binding protein C; mannose-binding protein C (liver); Mannose-binding protein C MBP-C Man-binding protein Ra-reactive factor polysaccharide-binding component p28A RaRF p28A Mbl2; MBP-C; RA-reactive factor P28A subunit; Ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A
Gene Aliases: Ab2-001; Ab2-011; L-MBP; MBL; MBL-C; Mbl2; MBP-C
UniProt ID: (Mouse) P41317, (Rat) P08661
Entrez Gene ID: (Mouse) 17195, (Rat) 64668
Molecular Function:
defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.