Invitrogen
Manufacturer Code:PA569379
Catalog # PIPA569379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human MBL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 27 kDa mannan-binding protein monomeric subunit; Collectin-1; mannan-binding lectin; mannan-binding lectin C; Mannan-binding protein; mannose binding lectin 2 (protein C); Mannose-binding lectin; mannose-binding lectin (protein C) 2, soluble; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding lectin C; mannose-binding lectin-C; Mannose-binding protein C; mannose-binding protein C (liver); MBL; MBL2; MBL2 man-binding lectin MBL MBP MBP1 COLEC1 HSMBPC mannose-binding lectin (protein C) 2 soluble (opsonic defect) man-binding protein mannose binding protein Mannose-binding lectin 2 soluble (opsonic defect) MGC116832 MGC116833 mannose-binding protein C soluble mannose-binding lectin; MBP-C; MBP1; pMBP-27; RA-reactive factor P28A subunit; Ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A; serum protein; MBL; soluble mannose-binding lectin
Gene Aliases: Ab2-001; Ab2-011; COLEC1; HSMBPC; MBL; MBL2; MBL2D; MBP; MBP-C; MBP1; MBPD; pMBP-27
UniProt ID: (Bovine) O02659, (Human) P11226, (Pig) Q9XSW3, (Rat) P08661
Entrez Gene ID: (Bovine) 281297, (Human) 4153, (Pig) 397230, (Goat) 102181921, (Horse) 100034103, (Rat) 64668
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.