Invitrogen
Manufacturer Code:PA546920
Catalog # PIPA546920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse MBL-2 Glu19-Asp244 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: COLEC1 HSMBPC MBL MBL2D MBP MBP-C MBP1 collectin-1 man-binding lectin mannose-binding lectin (protein C) 2 soluble (opsonic defect) mannose-binding lectin 2 soluble (opsonic defect) mannose-binding protein C; Mannan-binding protein; mannose binding lectin (C); mannose binding lectin, liver (C); Mannose-binding protein C; MBP-C; RA-reactive factor P28A subunit; RARF/P28A
Gene Aliases: L-MBP; MBL; MBL-C; Mbl2; MBP-C
UniProt ID: (Mouse) P41317
Entrez Gene ID: (Mouse) 17195
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.