Invitrogen
Manufacturer Code:PA5112057
Catalog # PIPA5112057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the human MBL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 27 kDa mannan-binding protein monomeric subunit Ab2-001 Ab2-011 BOS_23278 COLEC1 Collectin1 Collectin-1 HSMBPC L-MBP mannan-binding lectin Mannan-binding protein mannose binding lectin (C) mannose binding lectin 2 mannose binding lectin 2 (protein C) mannose binding lectin liver (C) Mannose-binding lectin mannose-binding lectin (protein C) 2 mannose-binding lectin (protein C) 2 soluble mannose-binding lectin (protein C) 2 soluble (opsonic defect) mannose-binding lectin 2 soluble (opsonic defect) mannose-binding lectin C Mannose-binding protein C mannose-binding protein C (liver) MBL Mbl2 MBL2D MBL-C MBP MBP1 MBPC MBP-C MBPD MGC116832 MGC116833 pMBP-27 RA-reactive factor P28A subunit Ra-reactive factor polysaccharide-binding component p28A RaRF p28A RARF/P28A soluble mannose-binding lectin; Collectin-1; mannan-binding lectin; Mannan-binding protein; Mannose-binding lectin; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); Mannose-binding protein C; MBP-C; MBP1
Gene Aliases: COLEC1; HSMBPC; MBL; MBL2; MBL2D; MBP; MBP-C; MBP1; MBPD
UniProt ID: (Human) P11226
Entrez Gene ID: (Human) 4153
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.