Invitrogen
Manufacturer Code:PA5109947
Catalog # PIPA5109947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 21-248 of human MBL2 (NP_000233.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 27 kDa mannan-binding protein monomeric subunit Ab2-001 Ab2-011 BOS_23278 COLEC1 Collectin1 Collectin-1 HSMBPC L-MBP Mannan-binding protein Mannose-binding lectin Mannose-binding protein C MBL Mbl2 MBL2D MBL-C MBP MBP1 MBPC MBP-C MBPD MGC116832 MGC116833 pMBP-27 RA-reactive factor P28A subunit Ra-reactive factor polysaccharide-binding component p28A RaRF p28A RARF/P28A; Collectin-1; mannan-binding lectin; Mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2 (protein C); mannose binding lectin, liver (C); Mannose-binding lectin; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); Mannose-binding protein C; mannose-binding protein C (liver); MBP-C; MBP1; RA-reactive factor P28A subunit; Ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A
Gene Aliases: Ab2-001; Ab2-011; COLEC1; HSMBPC; L-MBP; MBL; MBL-C; MBL2; MBL2D; MBP; MBP-C; MBP1; MBPD
UniProt ID: (Human) P11226, (Mouse) P41317, (Rat) P08661
Entrez Gene ID: (Human) 4153, (Mouse) 17195, (Rat) 64668
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.