Invitrogen
Manufacturer Code:MA533352
Catalog # PIMA533352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 8G6 |
Immunogen | Purified mouse MBL-A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Mannan-binding protein; mannose binding lectin (A); mannose binding lectin, serum (A); Mannose binding protein A MBP-A Ra-reactive factor polysaccharide-binding component p28B RaRF p28B MBL-1; Mannose-binding protein A; MBP-A; Ra-reactive factor polysaccharide-binding component p28B; RaRF p28B
Gene Aliases: MBL-A; Mbl1; MBP-A; S-MBP
UniProt ID: (Mouse) P39039
Entrez Gene ID: (Mouse) 17194
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.