Novus Biologicals
Manufacturer Code:NBP1601270.05MG
Catalog # NBP16012705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 8G6 |
Immunogen | mouse MBL-A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Mannan-binding protein; mannose binding lectin (A); mannose binding lectin, serum (A); Mannose-binding protein A; MBL-1 p28B RaRF p28B; MBP-A; Ra-reactive factor polysaccharide-binding component p28B; RaRF p28B
Gene Aliases: MBL-A; Mbl1; MBP-A; S-MBP
UniProt ID: (Mouse) P39039
Entrez Gene ID: (Mouse) 17194
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.