R&D Systems
Manufacturer Code:AF2077
Catalog # AF2077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse MBL-1 (R&D Systems Catalog # 2077-MB) Ser18-Ala239 Accession # P39039 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Mannan-binding protein; mannose binding lectin (A); mannose binding lectin, serum (A); Mannose-binding protein A; MBL1 MBL-1 MBL-A MBP-A p28B RaRF p28B S-MBP; MBP-A; Ra-reactive factor polysaccharide-binding component p28B; RaRF p28B
Gene Aliases: MBL-A; Mbl1; MBP-A; S-MBP
UniProt ID: (Mouse) P39039
Entrez Gene ID: (Mouse) 17194
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.