Novus Biologicals
Manufacturer Code:NBP272594C
Catalog # NBP272594C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A12 |
Immunogen | Full length recombinant protein of human MBL (NP_000233) produced in HEK293T cells. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: COLEC1collectin-1 Collectin-1 HSMBPC Mannan-binding protein Mannose-binding lectin mannose-binding lectin (protein C) 2 soluble mannose-binding lectin (protein C) 2 soluble (opsonic defect) mannose-binding protein C MBLmannan-binding lectin MBP MBP1mannose-binding lectin 2 soluble (opsonic defect) MBP-C MGC116832 MGC116833; Collectin-1; mannan-binding lectin; Mannan-binding protein; Mannose-binding lectin; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); Mannose-binding protein C; MBP-C; MBP1
Gene Aliases: COLEC1; HSMBPC; MBL; MBL2; MBL2D; MBP; MBP-C; MBP1; MBPD
UniProt ID: (Human) P11226
Entrez Gene ID: (Human) 4153
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.