Novus Biologicals
Manufacturer Code:NBP299368AF594
Catalog # NB085020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human MBD5. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1461 methyl-CpG binding domain protein 5 methyl-CpG-binding domain protein 5 methyl-CpG-binding protein MBD5 MRD1; Methyl-CpG-binding domain protein 5; Methyl-CpG-binding protein MBD5
Gene Aliases: KIAA1461; MBD5; MRD1
UniProt ID: (Human) Q9P267
Entrez Gene ID: (Human) 55777
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.