Novus Biologicals
Manufacturer Code:NBP224393
Catalog # NBP224393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 268-300 and 337-374 of human MBD4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3,N(4)-ethenocytosine glycosylase; EC 3.2.2.- G/T mismatch glycosylase G/U mismatch glycosylase MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics methyl-CpG binding domain protein 4 methyl-CpG-binding domain protein 43N(4)-ethenocytosine glycosylase Methyl-CpG-binding endonuclease 1 Methyl-CpG-binding protein MBD4 Mismatch-specific DNA N-glycosylase putative methyl-CpG binding protein; G/5-fluorouracil mismatch glycosylase with biphasic kinetics; G/T mismatch glycosylase; G/U mismatch glycosylase; methyl-CpG binding domain 4 DNA glycosylase; methyl-CpG binding domain protein 4; Methyl-CpG-binding domain protein 4; Methyl-CpG-binding endonuclease 1; Methyl-CpG-binding protein MBD4; Mismatch-specific DNA N-glycosylase; putative methyl-CpG binding protein
Gene Aliases: MBD4; MED1
UniProt ID: (Human) O95243
Entrez Gene ID: (Human) 8930
Molecular Function:
methyltransferase
nuclease
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.