Abnova
Manufacturer Code:PAB22578
Catalog # 89-124-289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MB21D1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MB21D1.
For Research Use Only
Protein Aliases: 2'3'-cGAMP synthase; cGAMP synthase; Cyclic GMP-AMP synthase; Mab-21 domain-containing protein 1; MGC131892; MGC142166; MGC142168; protein MB21D1
Gene Aliases: C6orf150; CGAS; h-cGAS; MB21D1
UniProt ID: (Human) Q8N884
Entrez Gene ID: (Human) 115004
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.