Invitrogen
Manufacturer Code:MA190116
Catalog # PIMA190116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | MECA-367 |
Immunogen | Balb/c mouse endothelial cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AV211525 MGC144710 MGC144711 MGC35998; MAdCAM-1; mMAdCAM-1; mucosal addressin cell adhesion molecule 1
Gene Aliases: AV211525
Entrez Gene ID: (Mouse) 17123
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.