Novus Biologicals
Manufacturer Code:NBP272568JF549
Catalog # N272568J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A5 |
Immunogen | Full length recombinant protein of human MAdCAM-1 (NP_570116) produced in HEK293T cells. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: hMAdCAM-1; hMAdCAM-1 MACAM1 MAdCAM-1 mucosal addressin cell adhesion molecule 1 mucosal addressin cell adhesion molecule-1 mucosal vascular addressin cell adhesion molecule 1; MAdCAM-1; Mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1
Gene Aliases: MACAM1; MADCAM1
UniProt ID: (Human) Q13477
Entrez Gene ID: (Human) 8174
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.