R&D Systems
Manufacturer Code:AF6056SP
Catalog # AF6056-SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MADCAM-1 Met1-Leu333 Accession # Q13477 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
Protein Aliases: hMAdCAM-1; hMAdCAM-1 MACAM1 MAdCAM1 MAdCAM-1 mucosal addressin cell adhesion molecule 1 mucosal addressin cell adhesion molecule-1 mucosal vascular addressin cell adhesion molecule 1; MAdCAM-1; Mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1
Gene Aliases: MACAM1; MADCAM1
UniProt ID: (Human) Q13477
Entrez Gene ID: (Human) 8174
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.