Invitrogen
Manufacturer Code:PA584160
Catalog # PIPA584160
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MAVS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARD adapter inducing interferon beta; CARD adaptor inducing IFN-beta; Cardif; Cardif IPS-1 KIAA1271 VISA; IFN-B promoter stimulator 1; Interferon beta promoter stimulator protein 1; IPS-1; MAVS; Mitochondrial antiviral-signaling protein; Putative NF-kappa-B-activating protein 031N; virus-induced signaling adaptor; Virus-induced-signaling adapter; VISA
Gene Aliases: CARDIF; IPS-1; IPS1; KIAA1271; MAVS; VISA
UniProt ID: (Human) Q7Z434
Entrez Gene ID: (Human) 57506
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.