Invitrogen
Manufacturer Code:PA520349
Catalog # PIPA520349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide from near the center of human VISA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARD adapter inducing interferon beta; CARD adaptor inducing IFN-beta; Cardif; CARDIF IPS-1 IPS1 VISA CARD adapter inducing interferon beta CARD adaptor inducing IFN-beta IFN-B promoter stimulator 1 interferon beta promoter stimulator protein 1 mitochondrial antiviral-signaling protein putative NF-kappa-B-activating protein 031N virus-induced signaling adaptor virus-induced-signaling adapter; IFN-B promoter stimulator 1; IFN-beta promoter stimulator-1; Interferon beta promoter stimulator protein 1; interferon-beta promoter stimulator protein 1; IPS-1; MAVS; mitochondrial anti-viral signaling protein; Mitochondrial antiviral-signaling protein; Putative NF-kappa-B-activating protein 031N; virus-induced signaling adapter; virus-induced signaling adaptor; Virus-induced-signaling adapter; VISA
Gene Aliases: CARDIF; D430028G21Rik; IPS-1; IPS1; KIAA1271; MAVS; VISA
UniProt ID: (Human) Q7Z434, (Mouse) Q8VCF0, (Rat) Q66HG9
Entrez Gene ID: (Human) 57506, (Mouse) 228607, (Rat) 311430
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.