Invitrogen
Manufacturer Code:PA520725
Catalog # PIPA520725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | a 12 amino acid peptide from near the amino terminus of human MATN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cartilage matrix protein; CMP CRTM cartilage matrix protein; matrilin 1, cartilage matrix protein 1; Matrilin-1
Gene Aliases: CMP; CRTM; Mat1; MATN1; matrilin-1
UniProt ID: (Human) P21941, (Mouse) P51942
Entrez Gene ID: (Human) 4146, (Rat) 297894, (Mouse) 17180
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.