Proteintech
Manufacturer Code:268731AP150UL
Catalog # 50-173-5483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MATE 2 Fusion Protein Ag25472 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hMATE 2 MATE 2 MATE2 MATE2 B MATE2 K SLC47A2; Kidney-specific H(+)/organic cation antiporter; MATE-2; Multidrug and toxin extrusion protein 2; solute carrier family 47 (multidrug and toxin extrusion), member 2; Solute carrier family 47 member 2
Gene Aliases: MATE2; MATE2-B; MATE2-K; MATE2K; SLC47A2
UniProt ID: (Human) Q86VL8
Entrez Gene ID: (Human) 146802, (Dog) 489527
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.