Novus Biologicals
Manufacturer Code:NBP128605
Catalog # NBP128605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A2 |
Immunogen | Recombinant human MAT2A (1-395aa) purified from E. coli (NP_005902). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AdoMet synthase 2; AdoMet synthase 2 AMS2 EC 2.5.1.6 MAT 2 MATA2SAMS2adoMet synthetase 2 MATII MAT-II Methionine adenosyltransferase 2 Methionine adenosyltransferase II methionine adenosyltransferase II alpha S-adenosylmethionine synthase isoform type-2 S-adenosylmethionine synthetase isoform type-2; adoMet synthetase 2; MAT 2; MAT-II; Methionine adenosyltransferase 2; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; S-adenosylmethionine synthase isoform type-2; testicular tissue protein Li 121
Gene Aliases: AMS2; MAT2A; MATA2; MATII; SAMS2
UniProt ID: (Human) A8K511
Entrez Gene ID: (Human) 4144
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.