Invitrogen
Manufacturer Code:PA572927
Catalog # PIPA572927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an N-terminal portion of human MAT2A (within residues 1-100). &lsqbSwiss-ProtNo. P31153] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AdoMet synthase 2; adoMet synthetase 2; MAT 2; MAT-II; MATA2 MATII SAMS2 MAT 2 MAT-II S-adenosylmethionine synthase isoform type-2 adoMet synthase 2 adoMet synthetase 2 methionine adenosyltransferase 2; Methionine adenosyltransferase 2; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; non-hepatic-type S-adenosylmethionine synthetase; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-2; testicular tissue protein Li 121
Gene Aliases: AMS2; D630045P18Rik; MAT2A; MATA2; MATII; SAMS2
UniProt ID: (Human) A8K511, (Rat) P18298, (Mouse) Q3THS6
Entrez Gene ID: (Human) 4144, (Bovine) 527403, (Rat) 171347, (Mouse) 232087
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.