Invitrogen
Manufacturer Code:PA5115550
Catalog # PIPA5115550
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAT2A corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AdoMet synthase 2; AdoMet synthase 2 adoMet synthetase 2 AMS2 D630045P18Rik MAT 2 Mat2a MATA2 MATII MAT-II Methionine adenosyltransferase 2 methionine adenosyltransferase 2A Methionine adenosyltransferase II methionine adenosyltransferase II alpha non-hepatic-type S-adenosylmethionine synthetase S-adenosylmethionine synthase isoform type-2 S-adenosylmethionine synthetase isoform type-2 SAMS2 testicular tissue protein Li 121; adoMet synthetase 2; MAT 2; MAT-II; Methionine adenosyltransferase 2; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; non-hepatic-type S-adenosylmethionine synthetase; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-2; testicular tissue protein Li 121
Gene Aliases: AMS2; D630045P18Rik; MAT2A; MATA2; MATII; SAMS2
UniProt ID: (Human) A8K511, (Rat) P18298, (Mouse) Q3THS6
Entrez Gene ID: (Human) 4144, (Rat) 171347, (Mouse) 232087
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.