Invitrogen
Manufacturer Code:PA541998
Catalog # PIPA541998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MAT1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AdoMet synthase 1; adoMet synthetase 1; MAT 1; MAT-I/III; Methionine adenosyltransferase 1; Methionine adenosyltransferase I alpha MAT MATA1 SAMS SAMS1; methionine adenosyltransferase I, alpha; Methionine adenosyltransferase I/III; S - adenosylmethionine synthetase; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthetase isoform type-1
Gene Aliases: AdoMet; AI046368; Ams; AMS1; MAT; MAT1A; MATA1; SADE; SAMS; SAMS1; SAS; wu:fi35e01; zgc:55442
UniProt ID: (Human) Q00266, (Bovine) Q2KJC6, (Rat) P13444, (Mouse) Q91X83
Entrez Gene ID: (Dog) 608788, (Rabbit) 100354729, (Horse) 100052797, (Human) 4143, (Bovine) 541078, (Rat) 25331, (Mouse) 11720, (Guinea pig) 100728681, (Zebrafish) 334195
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.