Invitrogen
Manufacturer Code:14579482
Catalog # 50-112-2694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 24B72D11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: achaete-scute complex homolog 1; achaete-scute complex homolog-like 1; achaete-scute complex-like protein 1; Achaete-scute homolog 1; ASH-1; KMT2H; mammalian achaete scute homolog 1; mASH-1
Gene Aliases: AI225900; Ascl1; ASH1; bHLHa46; Mash-1; Mash1
UniProt ID: (Rat) P19359, (Mouse) Q02067
Entrez Gene ID: (Rat) 64186, (Mouse) 17172
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.