Invitrogen
Manufacturer Code:PA5111828
Catalog # PIPA5111828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 212-224 of the rat Angiotensin-(1-7) Mas receptor (3rdintracellularloop) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: angiotensin-(1-7) receptor; c-mas Mas Mas1 Mas-1 Mas1 proto-oncogene MGC119966 MGRA Proto-oncogene Mas; MAS proto-oncogene; Mas-related G protein-coupled receptor A; MAS1 oncogene; Proto-oncogene Mas
Gene Aliases: c-mas; MAS; Mas-1; MAS1; MGRA
UniProt ID: (Human) P04201, (Rat) P12526, (Mouse) P30554
Entrez Gene ID: (Human) 4142, (Rat) 25153, (Mouse) 17171
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.