Invitrogen
Manufacturer Code:PA514993
Catalog # PIPA514993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from the C-terminal region of MARS2 conjugated to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: methionine tRNA ligase 2, mitochondrial; methionine--tRNA ligase 2; Methionine--tRNA ligase, mitochondrial; methionine-tRNA synthetase 2, mitochondrial; Methionyl-tRNA synthetase 2; Methionyl-tRNA synthetase mitochondrial &lsqbPrecursor] EC 6.1.1.10 Mitochondrial methionine--tRNA ligase MtMetRS Methionine--tRNA ligase 2 MARS2; methionyl-tRNA synthetase, mitochondrial; mitochondrial methionine--tRNA ligase; Mitochondrial methionyl-tRNA synthetase; MtMetRS
Gene Aliases: C730026E21Rik; COXPD25; MARS2; MetRS; mtMetRS
UniProt ID: (Human) Q96GW9, (Mouse) Q499X9
Entrez Gene ID: (Human) 92935, (Mouse) 212679
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.