Invitrogen
Manufacturer Code:PA578653
Catalog # PIPA578653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of mouse MARS. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: methionine tRNA ligase; Methionine--tRNA ligase, cytoplasmic; methionine-tRNA synthetase; Methionyl-tRNA synthetase; methionyl-tRNA synthetase, cytoplasmic; MetRS; Metrs Mtrns methionine-tRNA synthetase
Gene Aliases: Mars; Mars1; Metrs; Mtrns
UniProt ID: (Mouse) Q68FL6
Entrez Gene ID: (Rat) 299851, (Mouse) 216443
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.