Invitrogen
Manufacturer Code:PA540583
Catalog # PIPA540583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytosolic methionyl-tRNA synthetase; methionine tRNA ligase; Methionine--tRNA ligase, cytoplasmic; methionine-tRNA synthetase; Methionyl-tRNA synthetase; methionyl-tRNA synthetase, cytoplasmic; MetRS
Gene Aliases: CMT2U; id:ibd5106; ILFS2; ILLD; MARS; MARS1; METRS; MRS; MTRNS; SPG70; zgc:66122
UniProt ID: (Human) P56192, (Bovine) Q2T9L8, (Mouse) Q68FL6
Entrez Gene ID: (Human) 4141, (Rabbit) 100338022, (Bovine) 512531, (Dog) 474414, (Horse) 100053373, (Pig) 100516610, (Rat) 299851, (Mouse) 216443, (Guinea pig) 100721918, (Zebrafish) 338183
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.