Novus Biologicals
Manufacturer Code:NB11058875SS
Catalog # NB11058875T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length recombinant rat MARCKS expressed in and purified from E. coli. [UniProt# P30009] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 80K-L 80K-L protein FLJ14368 MACSmyristoylated alanine-rich C-kinase substrate MRACKS myristoylated alanine-rich protein kinase C substrate myristoylated alanine-rich protein kinase C substrate (MARCKS 80K-L) phosphomyristin PKCSLFLJ90045 PRKCSL Protein kinase C substrate 80 kDa protein light chain; 80K-L protein; MARCKS; Myristoylated alanine-rich C-kinase substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; Protein kinase C substrate, 80 kDa protein, light chain
Gene Aliases: 80K-L; MACS; MARCKS; PKCSL; PRKCSL
UniProt ID: (Human) P29966
Entrez Gene ID: (Human) 4082
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.